missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Catalase Reagent Dropper
Description
To perform tests, squeeze the dropper to crush the ampule. Invert the dropper for convenient drop-by-drop dispensing.
- Dropper contains one-day supply of reagent or stain, hermetically sealed inside an ampule
- Strict quality controls assure uniformity and accuracy
Specifications
Specifications
| Quantity | 50 Pack |
| Certifications/Compliance | CE |
| Includes | One day's worth of stain |
| Format | Dropper |
| For Use With (Application) | Used for determining the presence of catalase produced by bacteria |
| Storage Requirements | 15° to 30°C |
| Volume (Metric) | 0.5 mL |
| Product Type | Reagent/Stain Dropper |