missing translation for 'onlineSavingsMsg'
Learn More

BD 10% Potassium Hydroxide Reagent Dropper

Catalog No. 1491065
Change view
Click to view available options
Quantity:
50 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1491065 50 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1491065 Supplier BD Supplier No. 261191
Only null left

Identify microorganisms without mixing or measuring reagents

To perform tests, squeeze the dropper to crush the ampule. Invert the dropper for convenient drop-by-drop dispensing.

  • Dropper contains one-day supply of reagent or stain, hermetically sealed inside an ampule
  • Strict quality controls assure uniformity and accuracy

Specifications

Type Reagent/Stain Dropper
Quantity 50 Pack
Certifications/Compliance CE
Includes One day's worth of stain
Format Dropper
For Use With (Application) Used in the examination of direct smears for fungal element
Storage Requirements 15° to 30°C
Stain Type Potassium Hydroxide