Learn More
BD Plated Media M-ENDO Agar
Slightly selective and differential medium for isolation and differentiation of Enterobacteriaceae
Supplier: BD 298291
Description
- Used in enumeration of coliforms in water by the membrane filter technique
Specifications
| ISO/DIS, 11014 | |
| <30°C | |
| United States | |
| Plate |
| Purple | |
| Solid | |
| m Endo Agar | |
| 20/Pk. |