missing translation for 'onlineSavingsMsg'
Learn More

BD Nexiva IV Dual-Port Catheters

Catalog No. 22204416
Change view
Click to view available options
Needle Gauge:
18 G
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge
22204416 18 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22204416 Supplier BD Supplier No. 383539

Specifications

Material Vialon Biomaterial
Color Green
Needle Gauge 18 G
For Use With (Application) Designed to reduce insertion attempts, increase protection against blood exposure and needlestick injuries, simplify IV therapy process, enhance patient safety and satisfaction
Includes Flexible stabilization platform, Q-Syte™ luer access split septum dual port, 0.165cm I.D. extension tube
Latex Free Yes
Length (English) 1.25 in.
Length (Metric) 32 mm
Product Type Closed IV Catheter System
Disposable Single-use
Quantity 20/Pk.
Sterility Sterile
Show More Show Less