missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Microtainer™ Capillary Blood Collector and BD Microgard™ Closure
Textured inner surface and large tube diameter to aid sample mixing
Supplier: BD 365978
Description
- Designed to minimize sample contact and improve mixing
- “Twist assist” feature allows controlled closure removal; snap-lock fit assures complete seal
- Color-coded closures for accurate tube selection
- 8mm dia. for easy probe and pipet access; integral collector eliminates need for assembly
- Marked with fill lines, lot numbers and expiration date
- Holds 250 to 500μL of blood
- Extender (optional) facilitates labeling and handling
Chemical Identifiers
| Microgard | |
| 8 mm |
Specifications
| Amber | |
| Clot Activator and Gel | |
| 400 to 600 μL | |
| Yes |
| Serum Separator Tube | |
| Microgard | |
| 8 mm | |
| Amber |