missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Microtainer™ Capillary Blood Collector and BD Microgard™ Closure
Description
- Designed to minimize sample contact and improve mixing
- “Twist assist” feature allows controlled closure removal; snap-lock fit assures complete seal
- Color-coded closures for accurate tube selection
- 8mm dia. for easy probe and pipet access; integral collector eliminates need for assembly
- Marked with fill lines, lot numbers and expiration date
- Holds 250 to 500μL of blood
- Extender (optional) facilitates labeling and handling
Specifications
Specifications
| Product Type | Serum Tube |
| Additive | No Additive |
| Closure Type | Microgard |
| Capacity (Metric) | 250 to 500 μL |
| Diameter (Metric) Inner | 8 mm |
| Closure Color | Red |