Learn More
BD Macro-Vue™ RPR Cards and Components
Catalog No. B4971849
Change view
Click to view available options
Quantity:
10 Pack
300 Case
Type:
BD RPR Cards
Dispensing Needles, 20 Gauge
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
B4971849 300 Case BD RPR Cards
22130989 10 Pack Dispensing Needles, 20 Gauge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4971849 Supplier BD Supplier No. 271849
For large-scale testing in public health and clinical laboratories
RPR Card antigen suspension is a carbon particle cardiolipin antigen1 which detects reagin, an antibody-like substance present in serum or plasma from syphilitic persons, and occasionally in serum or plasma of persons with other acute or chronic conditions. The reagin binds to the test antigen, which consists of cardiolipin-lecithin-coated cholesterol particles, causing macroscopic flocculation.
  • CLIA Complexity: Moderate
  • Nontreponemal test
  • For the serological detection of syphilis as described in A Manual of Tests for Syphilis
Specifications
Detectable Analytes Reagin (Syphilis)
Type BD RPR Cards
For Use With (Application) For large-scale testing in public health and clinical laboratories
Includes BD Macro-Vue RPR cards, 18-mm circle, qualitative, 10 spot cards
Quantity 300 Case