Learn More
BD Luer-Lok™ PrecisionGlide™ Disposable Syringes with Detachable Needles: 3cc
Sterile, disposable syringes
$38.16 - $325.62
Specifications
| Material | Stainless Steel |
|---|---|
| Product Type | Syringe |
| Disposable | Single-use |
| Volume (Metric) | 3 mL |
| Sterility | Sterile |
| Catalog Number | Mfr. No. | Needle Gauge | Color | Graduations | Length (English) Needle | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Needle Gauge | Color | Graduations | Length (English) Needle | Price | Quantity | ||
|
14-823-37
|
BD
309571 |
23 G | Turquoise | 1/10 cc | 1 in. |
Pack of 100 Each for $40.70
Case of 8 Pack for $325.60
|
|
||
|
14-826-5E
|
BD
309580 |
18 G | Pink | 1/5 cc | 1.5 in. |
Pack of 100 Each for $40.70
Case of 8 Pack for $325.60
|
|
||
|
14-826-82
|
BD
309570 |
25 G | Blue | 1/10 cc | 0.62 in. |
Pack of 100 Each for $50.88
Case of 8 Pack for $407.04
|
|
||
|
14-826-84
|
BD
309577 |
21 G | Green | 1/10 cc | 1.5 in. |
Pack of 100 Each for $40.70
Case of 8 Pack for $325.60
|
|
||
|
14-829-13B
|
BD
309581 |
25 G | Blue | 1/10 cc | 1 in. |
Case of 800 Each for $325.62
|
|
||
|
14-829-13C
|
BD
309582 |
25 G | Blue | 1/10 cc | 1.5 in. |
Pack of 100 Each for $38.16
Case of 8 Pack for $305.28
|
|
||
|
14-826-5F
|
BD
309575 |
21 G | Green | 1/10 cc | 1 in. |
Pack of 100 Each for $38.16
Case of 8 Pack for $305.28
|
|
||
|
14-826-85
|
BD
309574 |
22 G | Black | 1/10 cc | 1.5 in. |
Pack of 100 Each for $39.54
Case of 8 Pack for $316.32
|
|
||
Description
Sterile, disposable syringes
- For easy identification of needle gauge, needle package and hub are color coded: 25G, blue; 24G, white; 23G, turquoise; 22G, black; 21G, green; 20G, yellow; 18G, pink
- All syringes are sealed in an Easy Peel™ package
- Autoclavable out of package
- With detachable PrecisionGlide™ needle
- Stainless-steel needle with polypropylene hub and Luer-Lok™ tip Cat. Nos. 14-826-11 and 14-826-7 are intramuscular needles with a thin-wall design
Specifications
| Stainless Steel | |
| Single-use | |
| Sterile |
| Syringe | |
| 3 mL |