Learn More
BD Intramedic™ Polyethylene Tubing
Sterile tubing
$636.05 - $636.05
| Catalog Number | Mfr. No. | Diameter (English) Inner | Diameter (English) Outer | Overall Length (English) | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (English) Inner | Diameter (English) Outer | Overall Length (English) | Price | Quantity | |||
|
22-270835
|
BD
427516 |
0.023 in. | 0.038 in. | 12 in. |
Pack of 12 Each for $636.05
|
|
|||
Description
- Option of PE 50 or PE 90
- Available in two lengths: 12 in. (30.5 cm) or 36 ft. (11m )
Specifications
| Polyethylene | |
| Sterile |