missing translation for 'onlineSavingsMsg'
Learn More

BD Intramedic™ Luer-Stub Adapters

Catalog No. p-3238367
Change view
Click to view available options
Size:
15 G
18 G
20 G
Color:
Orange
Pink
Yellow
For Use With (Equipment):
Intramedic™ PE100 and 90
Intramedic™ PE190 and 160
Intramedic™ PE240
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color For Use With (Equipment)
22044082 15 G Orange Intramedic™ PE240
1368121 18 G Pink Intramedic™ PE190 and 160
22044086 20 G Yellow Intramedic™ PE100 and 90
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22044082 Supplier BD Supplier No. B 427560

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fits Intramedic tubings

  • Sterile

Specifications

Size 15 G
Sterility Sterile
Color Orange
For Use With (Equipment) Intramedic™ PE240
Product Type Adapter