missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD GasPak™ EZ Gas Generating Systems and Supplies: Pouch Systems
Description
- One-step resealable pouches
- The pouches are specially designed to maximize the preservation of the desired environment throughout incubation
- This heat-free technology minimizes condensate, allowing excellent bacterial colony visualization though the pouch
Specifications
Specifications
| Type | EZ Anaerobe Pouch System |
| Closure Type | Ziplock |
| Format | Resealable Pouches |
| For Use With (Application) | Primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets |
| For Use With (Equipment) | BD GasPak EZ Small Incubation Container |
| Includes | Resealable pouches with indicator |
| Product Line | GasPak |
| Quantity | 20 Sachets |