missing translation for 'onlineSavingsMsg'
Learn More

BD GasPak™ EZ Gas Generating Systems and Supplies: Incubation Racks

Catalog No. B260673
Change view
Click to view available options
Type:
EZ Incubation Rack
Incubation Rack
Holds:
Petri Dishes (15 to 18)
Petri Dishes (30 to 33)
Petri Dishes (8 to 10)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Holds
B260673 EZ Incubation Rack Petri Dishes (15 to 18)
B260674 EZ Incubation Rack Petri Dishes (30 to 33)
B260003 Incubation Rack Petri Dishes (8 to 10)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B260673 Supplier BD Supplier No. 260673
Only null left

Catalyst-free convenience for use in producing anaerobic, microaerophilic, or CO2-enriched environments

Catalyst-free convenience for use in producing anaerobic, microaerophilic, or CO2-enriched environments

  • Holds 15-18 Petri dishes

Specifications

Type EZ Incubation Rack
Format Box Rack
For Use With (Application) Primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets
Holds Petri Dishes (15 to 18)
Product Line GasPak
Quantity Each
Size Standard