missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD DrySlide™ Reagent Slides
Description
- One-step procedure —no need for rehydrating
- No thin paper strips to manipulate and no glass slides are involved
- Each individual slide contains four reaction areas
- DrySlide Indole is a 30-second, spot indole test that uses DMABA (5% p-dimethylaminobenzaldehyde) formulation to test for indole-producing bacteria
- DrySlide Nitrocefin is a qualitative test to detect the presence of bacteria that produces β-lactamase and meets NCCLS guidelines; each slide contains a reagent cassette impregnated with nitrocefin
- DrySlide Oxidase with each slide is film-coated with an oxidase reagent (N, N, N',N'-tetramethyl-p-phenylenediamine dihydrochloride) to test for the presence of cytochrome C oxidase activity in bacteria; a positive result produces a purple color within 20 seconds
- DrySlide PYR Kit has L-pyrrolidonyl-β-naphthylamide that is used for the rapid presumptive identification of Group D Enterococci and Strep Group A; includes 15 slides and 15, 0.5mL crush ampules
Order Info
Slides are individually wrapped and supplied in convenient dispenser packs
Specifications
Specifications
| Quantity | 75 Pack |
| Certifications/Compliance | CE |
| Format | Slide |
| For Use With (Application) | Used for determining the oxidase reaction of bacteria |
| Storage Requirements | 15° to 30°C |
| Stain Type | Oxidase Reagent |
| Product Type | Oxidase Reagent |