missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Salmonella O QC Antigens

Catalog No. DF2130508
Change view
Click to view available options
Product Type:
Salmonella O Group A Antigen
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
DF2130508 Salmonella O Group A Antigen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog No. DF2130508 Supplier BD Supplier No. 221301
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Quick reactive Gold standard antisera-typing panels for Salmonella O antigen

  • Product type: Antigens and Antisera
  • Groupings: Q through S products
  • Sub type: Quality control antigens
  • Type: Antigens
  • Volume: 1mL
  • Shelf life: 730 days

Certifications

CE marked

Specifications

Product Type Salmonella O Group A Antigen
For Use With (Application) Used in the quality control of Salmonella O Antisera Poly B (225351), Factor 7 (228161), Group C1 Factors 6 (240985), and Poly A-I & Vi (240993)
Quantity 1 mL