missing translation for 'onlineSavingsMsg'
Learn More
Learn More
- Home
- Products
- Microbiology Quality Control Supplies
- Salmonella Antisera
- BD Difco™ Salmonella O Antisera and Quality Control Antigens
BD Difco™ Salmonella O Antisera and Quality Control Antigens
Catalog No. DF2815476
Change view
Click to view available options
Product Type:
Salmonella O Antiserum Factor 10, Group E1
Salmonella O Antiserum Factor 14
Salmonella O Antiserum Factor 15, Groups E2 and E3
Salmonella O Antiserum Factor 19, Group E4
Salmonella O Antiserum Factor 2
Salmonella O Antiserum Factor 20
Salmonella O Antiserum Factor 22, Group G1
Salmonella O Antiserum Factor 23, Group G2
Salmonella O Antiserum Factor 34, Group E3
Salmonella O Antiserum Factor 4, Group B
Salmonella O Antiserum Factor 5, Group B
Salmonella O Antiserum Factor 7, Groups C1 and C4
Salmonella O Antiserum Factor 8, Groups C2 and C3
Salmonella O Antiserum Factor 9, Group D
Salmonella O Antiserum Factors 4, 5, Group B
Salmonella O Antiserum Group D1 Factors 1, 9, 12
Salmonella O Antiserum Group E, Factors 1, 3, 10, 15, 19, 34
Salmonella O Antiserum Group E1 Factors 3, 10
Salmonella O Antiserum Group F Factor 11
Salmonella O Antiserum Group H Factors 1, 6, 14, 24, 25
Salmonella O Antiserum Poly A, Groups A, B, D, E1, E2, E3, E4 and L
Salmonella O Antiserum Poly B, Groups C1, C2, F, G and H
Salmonella O Antiserum Poly C, Groups I, J, K, M, N and O
Salmonella O Antiserum Poly D, Groups P, Q, R, S, T and U
Salmonella O Antiserum Poly E, Groups V, W, X, Y and Z
Salmonella O Antiserum Poly F, Groups 51-55
Salmonella O Antiserum Poly G, Groups 56-61
27 product options available for selection
| Catalog No. | Product Type |
|---|---|
| DF2815476 | Salmonella O Antiserum Factors 4, 5, Group B |
| B222621 | Salmonella O Antiserum Group H Factors 1, 6, 14, 24, 25 |
| DF2661471 | Salmonella O Antiserum Factor 14 |
| DF2951470 | Salmonella O Antiserum Group D1 Factors 1, 9, 12 |
| DF2257471 | Salmonella O Antiserum Factor 10, Group E1 |
| DF2258470 | Salmonella O Antiserum Factor 15, Groups E2 and E3 |
| DF2259479 | Salmonella O Antiserum Factor 19, Group E4 |
| DF2260476 | Salmonella O Antiserum Group F Factor 11 |
| DF2512472 | Salmonella O Antiserum Factor 34, Group E3 |
| DF2535475 | Salmonella O Antiserum Poly B, Groups C1, C2, F, G and H |
| DF2534476 | Salmonella O Antiserum Poly A, Groups A, B, D, E1, E2, E3, E4 and L |
| DF2536474 | Salmonella O Antiserum Poly C, Groups I, J, K, M, N and O |
| DF2537473 | Salmonella O Antiserum Poly D, Groups P, Q, R, S, T and U |
| DF2538472 | Salmonella O Antiserum Poly E, Groups V, W, X, Y and Z |
| DF2645472 | Salmonella O Antiserum Poly F, Groups 51-55 |
| DF2662470 | Salmonella O Antiserum Factor 20 |
| DF2659475 | Salmonella O Antiserum Factor 4, Group B |
| DF2646471 | Salmonella O Antiserum Poly G, Groups 56-61 |
| DF2660472 | Salmonella O Antiserum Factor 5, Group B |
| DF2664478 | Salmonella O Antiserum Factor 23, Group G2 |
| DF2663479 | Salmonella O Antiserum Factor 22, Group G1 |
| DF2814477 | Salmonella O Antiserum Factor 2 |
| DF2816475 | Salmonella O Antiserum Factor 7, Groups C1 and C4 |
| DF2817474 | Salmonella O Antiserum Factor 8, Groups C2 and C3 |
| DF2818473 | Salmonella O Antiserum Factor 9, Group D |
| DF2952479 | Salmonella O Antiserum Group E1 Factors 3, 10 |
| DF2819472 | Salmonella O Antiserum Group E, Factors 1, 3, 10, 15, 19, 34 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 27 options available.
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Description
Salmonella species cause a variety of human diseases called salmonelloses. The range of disease is from mild self-limiting gastroenteritis to more severe forms, possibly with bacteremia or typhoid fever, which can be life threatening. Hence diagnosis and identification of correct serotype is essential. These antisera aid in detecting such serotypes and the quality control antigens check their performance.
- Lyophilized, polyclonal rabbit antisera
- Polyvalent, specific for somatic antigens
- Cross-reactions are possible because serogroups may share non-major group antigens
- When properly rehydrated and used as recommended, each vial contains sufficient reagent for 60 tests
- For in vitro diagnostic use
- containing approximately 0.2% sodium azide as a preservative
Specifications
Specifications
| Certifications/Compliance | For in vitro diagnostic use |
| Product Type | Salmonella O Antiserum Factors 4, 5, Group B |