missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Salmonella O Antisera and Quality Control Antigens

Catalog No. DF2659475
Change view
Click to view available options
Product Type:
Salmonella O Antiserum Factor 10, Group E1
Salmonella O Antiserum Factor 14
Salmonella O Antiserum Factor 15, Groups E2 and E3
Salmonella O Antiserum Factor 19, Group E4
Salmonella O Antiserum Factor 2
Salmonella O Antiserum Factor 20
Salmonella O Antiserum Factor 22, Group G1
Salmonella O Antiserum Factor 23, Group G2
Salmonella O Antiserum Factor 34, Group E3
Salmonella O Antiserum Factor 4, Group B
Salmonella O Antiserum Factor 5, Group B
Salmonella O Antiserum Factor 7, Groups C1 and C4
Salmonella O Antiserum Factor 8, Groups C2 and C3
Salmonella O Antiserum Factor 9, Group D
Salmonella O Antiserum Factors 4, 5, Group B
Salmonella O Antiserum Group D1 Factors 1, 9, 12
Salmonella O Antiserum Group E, Factors 1, 3, 10, 15, 19, 34
Salmonella O Antiserum Group E1 Factors 3, 10
Salmonella O Antiserum Group F Factor 11
Salmonella O Antiserum Group H Factors 1, 6, 14, 24, 25
Salmonella O Antiserum Poly A, Groups A, B, D, E1, E2, E3, E4 and L
Salmonella O Antiserum Poly B, Groups C1, C2, F, G and H
Salmonella O Antiserum Poly C, Groups I, J, K, M, N and O
Salmonella O Antiserum Poly D, Groups P, Q, R, S, T and U
Salmonella O Antiserum Poly E, Groups V, W, X, Y and Z
Salmonella O Antiserum Poly F, Groups 51-55
Salmonella O Antiserum Poly G, Groups 56-61
27 product options available for selection
Product selection table with 27 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
DF2659475 Salmonella O Antiserum Factor 4, Group B
B222621 Salmonella O Antiserum Group H Factors 1, 6, 14, 24, 25
DF2661471 Salmonella O Antiserum Factor 14
DF2951470 Salmonella O Antiserum Group D1 Factors 1, 9, 12
DF2257471 Salmonella O Antiserum Factor 10, Group E1
DF2258470 Salmonella O Antiserum Factor 15, Groups E2 and E3
DF2259479 Salmonella O Antiserum Factor 19, Group E4
DF2260476 Salmonella O Antiserum Group F Factor 11
DF2512472 Salmonella O Antiserum Factor 34, Group E3
DF2535475 Salmonella O Antiserum Poly B, Groups C1, C2, F, G and H
DF2534476 Salmonella O Antiserum Poly A, Groups A, B, D, E1, E2, E3, E4 and L
DF2536474 Salmonella O Antiserum Poly C, Groups I, J, K, M, N and O
DF2537473 Salmonella O Antiserum Poly D, Groups P, Q, R, S, T and U
DF2538472 Salmonella O Antiserum Poly E, Groups V, W, X, Y and Z
DF2645472 Salmonella O Antiserum Poly F, Groups 51-55
DF2662470 Salmonella O Antiserum Factor 20
DF2646471 Salmonella O Antiserum Poly G, Groups 56-61
DF2660472 Salmonella O Antiserum Factor 5, Group B
DF2664478 Salmonella O Antiserum Factor 23, Group G2
DF2663479 Salmonella O Antiserum Factor 22, Group G1
DF2814477 Salmonella O Antiserum Factor 2
DF2815476 Salmonella O Antiserum Factors 4, 5, Group B
DF2816475 Salmonella O Antiserum Factor 7, Groups C1 and C4
DF2817474 Salmonella O Antiserum Factor 8, Groups C2 and C3
DF2818473 Salmonella O Antiserum Factor 9, Group D
DF2952479 Salmonella O Antiserum Group E1 Factors 3, 10
DF2819472 Salmonella O Antiserum Group E, Factors 1, 3, 10, 15, 19, 34
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 27 options available.
27 options
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog No. DF2659475 Supplier BD Supplier No. 226591
Edge
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Salmonella species cause a variety of human diseases called salmonelloses. The range of disease is from mild self-limiting gastroenteritis to more severe forms, possibly with bacteremia or typhoid fever, which can be life threatening. Hence diagnosis and identification of correct serotype is essential. These antisera aid in detecting such serotypes and the quality control antigens check their performance.

  • Lyophilized, polyclonal rabbit antisera
  • Polyvalent, specific for somatic antigens
  • Cross-reactions are possible because serogroups may share non-major group antigens
  • When properly rehydrated and used as recommended, each vial contains sufficient reagent for 60 tests
  • For in vitro diagnostic use
  • containing approximately 0.2% sodium azide as a preservative

Specifications

Certifications/Compliance For in vitro diagnostic use
Product Type Salmonella O Antiserum Factor 4, Group B