Learn More
BD Difco™ Salmonella O Antisera and Quality Control Antigens
Supplier:  BD 225341
 View more versions of this product
Catalog No. DF2534476
Only null left
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Description
Description
Salmonella species cause a variety of human diseases called salmonelloses. The range of disease is from mild self-limiting gastroenteritis to more severe forms, possibly with bacteremia or typhoid fever, which can be life threatening. Hence diagnosis and identification of correct serotype is essential. These antisera aid in detecting such serotypes and the quality control antigens check their performance.
  • Lyophilized, polyclonal rabbit antisera
  • Polyvalent, specific for somatic antigens
  • Cross-reactions are possible because serogroups may share non-major group antigens
  • When properly rehydrated and used as recommended, each vial contains sufficient reagent for 60 tests
  • For in vitro diagnostic use
  • containing approximately 0.2% sodium azide as a preservative
Specifications
Chemical Identifiers
For in vitro diagnostic use
Specifications
For in vitro diagnostic use
Salmonella O Antiserum Poly A, Groups A, B, D, E1, E2, E3, E4 and L
Product Suggestions
Product Suggestions