missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ YPD Agar
Description
General methods in yeast genetics specify using yeast extract-peptone-dextrose (YPD) medium for cultivating Saccharomyces cerevisiae and other yeasts. Yeasts grow well on a minimal medium containing only dextrose and salts. The addition of protein and yeast cell extract hydrolysates allows faster growth so that during exponential or log-phase growth, the cells divide every 90 minutes.
- Peptone acts as a source of carbon, nitrogen, vitamins and minerals
- Yeast extract supplies B-complex vitamins, which stimulate bacterial growth
- Dextrose is the carbohydrate source
- Agar acts as the solidifying agent
- pH (6.5% solution, 25°C): 6.5 ± 0.2
Order Info
Prices are for units of one each unless otherwise specified.
Specifications
Specifications
| Color | Beige |
| Content And Storage | <30°C |
| Form | Dehydrated Powder |
| Media Type | YPD Agar |
| Product Type | Nutrient Agar |
| Formulation | Yeast extract 10g/L, Peptone 20g/L, Dextrose 20g/L, Agar 15g/L |
| For Use With (Application) | For maintaining and propagating Yeasts in molecular microbiology procedures |
| Packaging Type | Bottle |
| Quantity | 500 g |