missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Yeast Mold Agar

Catalog No. df0712170
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0712170 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0712170 Supplier BD Supplier No. 271210

Used for cultivating yeasts, molds and other aciduric microorganisms. BD™ Difco™ Yeast Mold Agar is prepared according to the formulae published by Wickerham. Supplied as a dehydrated powder for ease of use.

Media selectivity may be enhanced through acidification or through addition of selective agents. YM Agar should be sterilized without pH adjustment and sterile acid added to the sterile molten medium cooled to 45–50°C. Acidified YM Agar should not be heated. Antibiotics may be aseptically added to the sterile media. Other fungistatic materials, such as sodium propionate and diphenyl may be added to YM Agar to eliminate molds and permit the enumeration of yeasts in mixed populations.

  • Good cultural response at pH 6.2 (4.1% solution, 25°C):
    Aspergillus brasiliensis (niger) 16404, Candida albicans 10231, Escherichia coli 25922, Lactobacillus rhamnosus 7469, Saccharomyces cerevisiae 9763
  • Yeast extract is a source of trace elements, vitamins and amino acids
  • Malt extract is a source of carbon, protein and nutrients
  • Peptone is an additional source of carbon and provides nitrogen and amino acids
  • Dextrose provides carbon
  • Agar is the solidifying agent

Order Info

Prices are for units of one each unless otherwise specified.

Specifications

Certifications/Compliance AOAC, COMPF
Color Beige
Content And Storage <30°C
Form Dehydrated Powder
Media Type Yeast Mold Agar
Product Type Agar
Formulation Yeast extract 3g/L, Malt extract 3g/L, Peptone 5g/L, Dextrose 10g/L, Agar 20g/L
For Use With (Application) Cultivation of yeasts, molds and other aciduric microorganisms
Packaging Type Bottle
Quantity 500 g