missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Xylose Lysine Deoxycholate (XLD) Agar

Catalog No. DF0788179
Change view
Click to view available options
Quantity:
500 g
Packaging Type:
Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
DF0788179 500 g Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0788179 Supplier BD Supplier No. 278850
Edge

Moderately selective medium recommended for isolation and differentiation of enteric pathogens, especially Shigella species

  • Formulation: Xylose 3.5g/L, L-Lysine 5g/L, Lactose 7.5g/L, Saccharose 7.5g/L, Sodium Chloride 5g/L, Yeast extract 3g/L, Phenol red 0.08g/L, Sodium Desoxycholate 2.5g/L, Ferric Ammonium Citrate 0.8g/L, Sodium Thiosulfate 6.8g/L, Agar 13.5g/L

Order Info

Prices are for units of one each unless otherwise specified.

Certifications

AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, ISO, JP, MCM9, SMD, SMWW, USP

Specifications

Certifications/Compliance AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, ISO, JP, MCM9, SMD, SMWW, USP
Content And Storage <30°C
Form Dehydrated
Media Type XLD Agar
Product Type Nutrient Agar
Formulation Xylose 3.5g/L, L-Lysine 5g/L, Lactose 7.5g/L, Saccharose 7.5g/L, Sodium Chloride 5g/L, Yeast extract 3g/L, Phenol red 0.08g/L, Sodium Desoxycholate 2.5g/L, Ferric Ammonium Citrate 0.8g/L, Sodium Thiosulfate 6.8g/L, Agar 13.5g/L
For Use With (Application) Isolation and differentiation of enteric pathogens
Packaging Type Bottle
Quantity 500 g