missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Wallerstein Laboratory Nutrient Broth
Description
- Formulation: Yeast extract 4g/L, Pancreatic digest of Casein 5g/L, Dextrose 50g/L, Monopotassium Phosphate 0.55g/L, Potassium Chloride 425mg/L, Calcium Chloride 125mg/L, Magnesium Sulfate 125mg/L, Ferric Chloride 2.5mg/L, Manganese Sulfate 2.5mg/L, Bromcresol Green 22mg
Order Info
Prices are for units of one each unless otherwise specified.
Certifications
ISO/DIS, 11014
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Nutrient Broth |
| Product Type | Microbiology Media |
| Formulation | Yeast extract 4g/L, Pancreatic digest of Casein 5g/L, Dextrose 50g/L, Monopotassium Phosphate 0.55g/L, Potassium Chloride 425mg/L, Calcium Chloride 125mg/L, Magnesium Sulfate 125mg/L, Ferric Chloride 2.5mg/L, Manganese Sulfate 2.5mg/L, Bromcresol Green 22mg |
| For Use With (Application) | Cultivating yeasts, molds and bacteria encountered in brewing and industrial fermentation processes |
| Packaging Type | Bottle |
| Quantity | 500 g |