missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Violet Red Bile Glucose Agar

Catalog No. DF1866172
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF1866172 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF1866172 Supplier BD Supplier No. 218661
Edge

For detecting and enumerating Enterobacteriaceae in food and dairy products

  • Formulation: Yeast extract 3g/L, Pancreatic digest of Gelatin 7g/L, Bile Salts No. 3 1.5g/L, Glucose 10g/L, Sodium Chloride 5g/L, Neutral red 0.03g/L, Crystal Violet 2g/L, Agar 15g/L

Order Info

Prices are for units of one each unless otherwise specified

Certifications

CCAM, COMPF, EP, ISO, JP, USP

Specifications

Certifications/Compliance CCAM, COMPF, EP, ISO, JP, USP
Color Violet Red
Content And Storage 2°C to 30°C
Form Dehydrated
Media Type Violet Red Bile Glucose Agar
Product Type Agar
Formulation Yeast extract 3g/L, Pancreatic digest of Gelatin 7g/L, Bile Salts No. 3 1.5g/L, Glucose 10g/L, Sodium Chloride 5g/L, Neutral red 0.03g/L, Crystal Violet 2g/L, Agar 15g/L
For Use With (Application) For detecting and enumerating Enterobacteriaceae in food and dairy products
Packaging Type Bottle
Quantity 500 g