missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Microbial Content Test Agar
Recommended for the detection and enumeration of microorganisms present on surfaces of sanitary importance
$436.68 - $1,397.68
Specifications
| Certifications/Compliance | CCAM |
|---|---|
| Form | Dehydrated |
| Media Type | Microbial Content Test Agar |
| For Use With (Application) | Detection and enumeration of microorganisms |
| Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity | ||||
|
DF0553-17-2
|
BD
255320 |
500 g | Bottle |
N/A
|
|
||||
|
DF0553-07-4
|
BD
255310 |
2 kg | Pail |
N/A
|
|
||||
Description
Certifications
CCAM
Specifications
| CCAM | |
| Dehydrated | |
| Microbial Content Test Agar | |
| Detection and enumeration of microorganisms |