missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Microbial Content Test Agar

Catalog No. DF0553172
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Bottle
Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
DF0553172 500 g Bottle
DF0553074 2 kg Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. DF0553172 Supplier BD Supplier No. 255320
Edge

Recommended for the detection and enumeration of microorganisms present on surfaces of sanitary importance

Certifications

CCAM

Specifications

Certifications/Compliance CCAM
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Microbial Content Test Agar
For Use With (Application) Detection and enumeration of microorganisms
Packaging Type Bottle
Quantity 500 g