missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: TCBS Agar Thiosulfate-Citrate-Bile-Sucrose Agar

Catalog No. DF0650174
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0650174 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0650174 Supplier BD Supplier No. 265020

For selective isolation and cultivation of V. cholerae and other enteropathogenic vibrios

TCBS is the primary plating medium universally used for the selective isolation of vibrios that cause cholera, diarrhea and food poisoning. It was developed by Kobayashi et al.4, who modified the selective medium of Nakanishi.5 The combination
of alkaline peptone water and TCBS Agar is used in many procedures for the isolation of V. cholerae and other Vibrio species from feces.

Specifications

Certifications/Compliance ISO/DIS, 11014, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, ISO, MCM9, SMWW
Content And Storage <30°C
Form Dehydrated
Media Type Thiosulfate Citrate Bile Salts Sucrose (TCBS) Agar
Product Type Agar
Formulation Yeast extract 5g/L, Proteose Peptone No. 3 10g/L, Sodium Citrate 10g/L, Sodium Thiosulfate 10g/L, Oxgall 8g/L, Saccharose 20g/L, Sodium Chloride 10g/L, Ferric Ammonium Citrate 1g/L, Bromthymol blue 0.04g/L, Thymol blue 0.04g/L, Agar 15g/L
For Use With (Application) Isolation of Cholera vibrios and Vibrio parahaemolyticus
Packaging Type Bottle
Quantity 500 g