missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: TAT Broth Base

Catalog No. DF0984171
Change view
Click to view available options
Quantity:
500 g
2000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0984171 500 g
1491097 2000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. DF0984171 Supplier BD Supplier No. 298410
Edge

For sterility testing of viscous materials, adapted to sterility of cosmetics

Certifications

ISO/DIS, 11014

Specifications

Content And Storage <30°C
Form Dehydrated
Media Type TAT Broth Base
Product Type Nutrient Broth
Formulation Pancreatic digest of Casein 20g/960mL, Soy Lecithin 5g/960mL
For Use With (Application) Microbial growth
Packaging Type Bottle
Quantity 500 g