missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Purple Broth Base

Catalog No. DF0227178
Change view
Click to view available options
Content And Storage:
<30°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Content And Storage
DF0227178 <30°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0227178 Supplier BD Supplier No. 222710
Edge

Basal medium to which carbohydrates are added to determine fermentation reactions

Specifications

Content And Storage <30°C
Form Dehydrated
Media Type Purple Broth Base
Product Type Microbiology Media
Formulation Pancreatic digest of Gelatin 10g/L, Sodium Chloride 5g/L, Bromcresol Purple 0.02g/L
For Use With (Application) Basal medium to which carbohydrates are added to determine fermentation reactions
Packaging Type Bottle
Quantity 500 g