missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Potato Dextrose Broth

Catalog No. DF0549179
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0549179 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0549179 Supplier BD Supplier No. B254920
Edge

For cultivation of Yeasts and mold. BD Difco™ Dehydrated Culture Media: Potato Dextrose Broth is part of a large selection of dehydrated media.

  • Dehydrated
  • Culture Media
  • Potato Dextrose

Specifications

Certifications/Compliance CE, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, JP, MCM9, SMD, USP
Content And Storage Refrigerate
Form Dehydrated
Media Type Potato Dextrose Broth
Product Type Microbiology Media
Formulation Potato Starch (from infusion) 4g/L, Dextrose 20g/L
For Use With (Application) Cultivation of Yeasts and mold
Packaging Type Bottle
Quantity 500 g