missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Mueller Hinton Broth

Catalog No. DF0757176
Change view
Click to view available options
Quantity:
500 g
Packaging Type:
Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
DF0757176 500 g Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0757176 Supplier BD Supplier No. 275730

Used in the cultivation of a wide variety of fastidious and nonfastidious microorganisms. BD™ Difco™ Mueller Hinton Broth is a general purpose medium that is not cation adjusted. It may be used in antimicrobial susceptibility testing of microorganisms by broth dilution methods.

Mueller Hinton Broth, not cation-adjusted, is a dehydrated culture medium that is not supplemented with calcium or magnesium ions. It is formulated to have a low thymine and thymidine content. It may be used for broth dilution antimicrobial susceptibility testing as long as the calcium and magnesium ion concentrations are adjusted according to CLSI standard M7.

  • Acid hydrolysate (digest) of casein and beef extract supply amino acids and other nitrogenous substances, minerals, vitamins, carbon and other nutrients to support the growth of microorganisms
  • Starch acts as a protective colloid against toxic substances in the medium; hydrolysis of the starch during autoclaving provides a small amount of dextrose, which is a source of energy
  • Displays a cultural response to: Enterococcus faecalis 29212, Escherichia coli 25922, Pseudomonas aeruginosa 27853 and Staphylococcus aureus 29213
  • pH (2.1% solution, 25°C): 7.3 ± 0.1
  • Calcium: 2.9–5.9mg/L; Magnesium: 3.2–5.2mg/L

Specifications

Certifications/Compliance CE, CLSI
Color Light beige (with a few dark specks)
Content And Storage Refrigerate
Form Dehydrated Powder
Media Type Mueller Hinton Broth
Product Type Nutrient Broth
Formulation Beef extract 2g/L, Acid digest of Casein 17.5g/L, Starch 1.5g/L
For Use With (Application) Cultivation of wide variety of fastidious and non-fastidious microorganisms
Packaging Type Bottle
Quantity 500 g