missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Modified Letheen Broth (AOAC)

Catalog No. DF0630170
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0630170 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0630170 Supplier BD Supplier No. 263010
Edge

For determining antibiotic potency by the microbiological assay technique

Applications:

For evaluation, maintenance, selective enrichment, differentiation, isolation, cultivation, detection, propagation of bacteria

Specifications

Certifications/Compliance BAM
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Letheen Broth
Product Type Microbiology Media
Formulation Letheen broth 25.7g/L, Tryptone 5g/L, Proteose Peptone No. 3 10g/L, Yeast extract 2g/L, Sodium Bisulfite 0.1g/L
For Use With (Application) Microbiological evaluation of cosmetics - inactivates preservative agents
Packaging Type Bottle
Quantity 500 g