missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: mFC Broth Base
For cultivating and enumerating fecal Coliforms by the membrane filter technique at elevated temperature
Supplier: BD 288330
Description
- Use with Rosolic Acid
- Formulation: Tryptose 10g/L, Proteose Peptone 5g/L, Yeast extract 3g/L, Lactose 12.5g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Aniline blue 0.1g/L
Certifications
CE, EPA, SMWW
Specifications
| CE, EPA, SMWW | |
| Dehydrated | |
| Microbiology Media | |
| Used with Rosolic Acid in cultivating and enumerating fecal Coliforms by the membrane filter technique at elevated temperature | |
| 500 g |
| Refrigerate | |
| mFC Broth Base | |
| Tryptose 10g/L, Proteose Peptone 5g/L, Yeast extract 3g/L, Lactose 12.5g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Aniline blue 0.1g/L | |
| Bottle |