missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Marine Agar 2216

Catalog No. DF0979178
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0979178 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0979178 Supplier BD Supplier No. 212185
Edge

Isolation, cultivation and enumeration of heterotrophic marine bacteria. BD Difco™ Dehydrated Culture Media: Marine Agar 2216 is part of a large selection of dehydrated media.

  • Dehydrated
  • Culture Media
  • Marine Agar 2216

Order Info

See our web site or the Fisher Scientific™ HealthCare catalog for product information.

Specifications

Content And Storage <30°C
Form Dehydrated
Media Type Marine Agar 2216
Product Type Agar
Formulation Peptone 5g/L, Yeast extract 1g/L, Ferric Citrate 0.1g/L, Sodium Chloride 19.45g/L, Magnesium Chloride 8.8g/L, Sodium Sulfate 3.24g/L, Calcium Chloride 1.8g/L, Potassium Chloride 0.55g/L, Sodium Bicarbonate 0.16g/L, Potassium Bromide 0.08g/L, Strontium Chloride 34mg/L, Boric Acid 22mg/L, Sodium Silicate 4mg/L, Sodium Fluoride 2.4mg/L, Ammonium Nitrate 1.6mg/L, Disodium Phosphate 8mg/L, Agar 15g/L
For Use With (Application) Isolation, cultivation and enumeration of heterotrophic marine bacteria
Packaging Type Bottle
Quantity 500 g