missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ MacConkey Broth

Catalog No. DF0020177
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0020177 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0020177 Supplier Fisherbrand™ Supplier No. B220100
Edge

A selective medium for detecting gram-negative, lactose-fermenting coliforms. Fisherbrand™ MacConkey Broth is provided dehydrated.

  • 500g bottle
  • Works excellently with milk or water

Order Info

See our web site or the Fisher Scientific™ HealthCare catalog for product information.

Specifications

Certifications/Compliance CE, EP, ISO, JP, USP
Content And Storage Refrigerate
Form Dehydrated
Media Type MacConkey Broth
Product Type Microbiology Media
Formulation Oxgal 5g/L, Pancreatic digest of Gelatin 20g/L, Lactose 10g/L, Bromcresol Purple 0.01g/L
For Use With (Application) Detection of Coliform organsms in Milk and Water
Packaging Type Bottle
Quantity 500 g