missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: MacConkey Agar Base
Description
- Format: Dehydrated
Order Info
See our web site or the Fisher Scientific™ HealthCare catalog for product information.
Specifications
Specifications
| Certifications/Compliance | ISO 9001:2008, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, JP, MCM9, SMD, SMWW, USP |
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | MacConkey Agar |
| Product Type | Agar |
| Formulation | Pancreatic digest of Gelatin 3g/L, Peptones (meat and Casein) 3g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Neutral red 0.03g/L, Crystal Violet 1mg/L |
| For Use With (Application) | Added Carbohydrate in differentiating Coliforms based on fermentation reaction |
| Packaging Type | Bottle |
| Quantity | 500 g |