missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: m LES Endo Agar

Catalog No. DF0736172
Change view
Click to view available options
Quantity:
500 g
For Use With (Application):
Coliform enumeration
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity For Use With (Application)
DF0736172 500 g Coliform enumeration
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0736172 Supplier BD Supplier No. 273620
Edge

For enumerating coliforms in water by membrane filtration

  • Formulation: Yeast extract 1.2g/L, Casitone 3.7g/L, Thiopeptone 3.7g/L, Tryptose 7.5g/L, Lactose 9.4g/L, Dipotassium Phosphate 3.3g/L, Monopotassium Phosphate 1g/L, Sodium Chloride 3.7g/L, Sodium Desoxycholate 0.1g/L, Sodium Lauryl Sulfate 0.05g/L, Sodium Sulfite 1.6g

Certifications

CE, COMPF, SMD, SMWW

Specifications

Certifications/Compliance CE, COMPF, SMD, SMWW
Content And Storage <30°C
Form Dehydrated
Media Type mLES Endo Agar
Product Type Agar
Formulation Yeast extract 1.2g/L, Casitone 3.7g/L, Thiopeptone 3.7g/L, Tryptose 7.5g/L, Lactose 9.4g/L, Dipotassium Phosphate 3.3g/L, Monopotassium Phosphate 1g/L, Sodium Chloride 3.7g/L, Sodium Desoxycholate 0.1g/L, Sodium Lauryl Sulfate 0.05g/L, Sodium Sulfite 1.6g/L, Basic Fuchsin 0.8g/L, Agar 15g/L
For Use With (Application) Coliform enumeration
Packaging Type Bottle
Quantity 500 g