missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Listeria Enrichment Broth
Description
For evaluation, maintenance, selective enrichment, differentiation, isolation, cultivation, detection, propagation of bacteria
Specifications
Specifications
| Certifications/Compliance | CE |
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | Listeria Enrichment Broth |
| Product Type | Microbiology Media |
| Formulation | Pancreatic digest of Casein 17g/L, Soytone 3g/L, Dextrose 2.5g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Yeast extract 6g/L, Cycloheximide 0.05g/L, Acriflavine HCl 15mg/L, Nalidixic Acid 0.04g/L |
| For Use With (Application) | Selectively enrich Listeria from food |
| Packaging Type | Bottle |
| Quantity | 500 g |