missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: LB Broth, Miller (Luria-Bertani)

Catalog No. DF0446075
Change view
Click to view available options
Quantity:
500 g
2000 g
Packaging Type:
Bottle
Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
DF0446075 2000 g Pail
DF0446173 500 g Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. DF0446075 Supplier BD Supplier No. B244610

Propagate and maintain E. coli for molecular biology. BD Difco™ Dehydrated Culture Media: LB Broth, Miller (Luria-Bertani) supplied in dehydrated form for the convenience of the laboratorian and bioprocessing specialist.

 

  • Contains 10g/L NaCl
  • For propagation of E. coli
  • Wide range of molecular biology applications

 

Specifications

Certifications/Compliance CCAM
Content And Storage Refrigerate
Form Dehydrated
Media Type LB Broth, Miller
Product Type Microbiology Media
Formulation Tryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L
For Use With (Application) Propagation and maintenance of Escherichia coli for molecular biology
Packaging Type Pail
Quantity 2000 g