missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Lauryl Tryptose Broth

Catalog No. DF0241170
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Bottle
Pail
Certifications/Compliance:
AOAC, BAM, CCAM, COMPF, ISO, SMD, SMWW
CE, AOAC, BAM, CCAM, COMPF, ISO, SMD, SMWW
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type Certifications/Compliance
DF0241170 500 g Bottle CE, AOAC, BAM, CCAM, COMPF, ISO, SMD, SMWW
DF0241072 2 kg Pail AOAC, BAM, CCAM, COMPF, ISO, SMD, SMWW
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. DF0241170 Supplier BD Supplier No. 224150

Medium for detection of coliform bacteria

Specifications

Certifications/Compliance CE, AOAC, BAM, CCAM, COMPF, ISO, SMD, SMWW
Content And Storage Refrigerate
Form Dehydrated
Media Type Lauryl Tryptose Broth
Product Type Nutrient Broth
Formulation Tryptose 20g/L, Lactose 5g/L, Dipotassium Phosphate 2.75g/L, Monopotassium Phosphate 2.75g/L, Sodium Chloride 5g/L, Sodium Lauryl Sulfate 0.1g/L
For Use With (Application) Detection of Coliform organisms in materials of sanitary importance
Packaging Type Bottle
Quantity 500 g