missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Lactobacilli MRS Agar

Catalog No. DF0882170
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0882170 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0882170 Supplier BD Supplier No. 288210
Edge

For cultivation of lactobacilli. BD Difco™ Dehydrated Culture Media: Lactobacilli MRS Agar is part of a large selection of dehydrated media.

  • Dehydrated
  • Culture Media
  • Lactobacilli MRS Agar

Specifications

Certifications/Compliance COMPF, ISO, SMD
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type MRS Agar
Product Type Agar
Formulation Proteose Peptone No. 3 10g/L, Beef extract 10g/L, Yeast extract 5g/L, Dextrose 20g/L, Polysorbate 80 1g/L, Ammonium Citrate 2g/L, Sodium Acetate 5g/L, Magnesium Sulfate 0.1g/L, Manganese Sulfate 0.05g/L, Dipotassium Phosphate 2g/L, Agar 15g/L
For Use With (Application) Cultivation of lactobacilli
Packaging Type Bottle
Quantity 500 g