missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Fraser Broth Base

Catalog No. DF0219070
Change view
Click to view available options
Quantity:
500 g
2000 g
Packaging Type:
Bottle
Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
DF0219070 2000 g Pail
DF0219178 500 g Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. DF0219070 Supplier BD Supplier No. 211766

Use with Fraser Broth Supplement Selective enrichment and detection of Listeria

  • Formulation: Pancreatic digest of Casein 5g/L, Proteose Peptone No.3 5g/L, Beef extract 5g/L, Yeast extract 5g/L, Sodium Chloride 20g/L, Disodium Phosphate 9.6g/L, Monopotassium Phosphate 1.35g/L, Esculin 1g/L, Nalidixic Acid 0.02g/L, Acriflavine HCl 24mg/L, Lithium Chloride 3g/L

Certifications

CCAM, COMPF, ISO, USDA

Specifications

Certifications/Compliance CCAM, COMPF, ISO, USDA
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Fraser Broth Base
Product Type Nutrient Broth
Formulation Pancreatic digest of Casein 5g/L, Proteose Peptone No.3 5g/L, Beef extract 5g/L, Yeast extract 5g/L, Sodium Chloride 20g/L, Disodium Phosphate 9.6g/L, Monopotassium Phosphate 1.35g/L, Esculin 1g/L, Nalidixic Acid 0.02g/L, Acriflavine HCl 24mg/L, Lithium Chloride 3g/L
For Use With (Application) Used with Fraser broth supplement selective enrichment and detection of Listeria
Packaging Type Pail
Quantity 2000 g