missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: EE Broth Mossel Enrichment

Catalog No. DF0566177
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0566177 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0566177 Supplier BD Supplier No. 256620
Edge

For selectively enriching and detecting Enterobacteriaceae, particularly from foods

  • Formulation: Pancreatic digest of Gelatin 10g/L, Dextrose 5g/L, Disodium Phosphate 8g/L, Monopotassium Phosphate 2g/L, Brilliant green 15mg/L, Oxgall 20g/mL

Certifications

CE, COMPF, EP, ISO, JP, USP

Specifications

Certifications/Compliance CE, COMPF, EP, ISO, JP, USP
Content And Storage <30°C
Form Dehydrated
Media Type EE Broth
Product Type Microbiology Media
Formulation Pancreatic digest of Gelatin 10g/L, Dextrose 5g/L, Disodium Phosphate 8g/L, Monopotassium Phosphate 2g/L, Brilliant green 15mg/L, Oxgall 20g/mL
For Use With (Application) For selectively enriching and detecting Enterobacteriaceae, particularly from food
Packaging Type Bottle
Quantity 500 g