missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Dey/Engley (D/E) Neutralizing Broth

Catalog No. DF0819172
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0819172 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0819172 Supplier BD Supplier No. 281910
Edge

Tests efficiency of disinfectants and sanitation. BD Difco™ Dehydrated Culture Media: Dey/Engley (D/E) Neutralizing Broth is part of a large selection of dehydrated media.

  • Dehydrated
  • Culture Media
  • Dey/Engley (D/E) Neutralizing Broth

Specifications

Certifications/Compliance AOAC, BAM, USDA
Content And Storage Refrigerate
Form Dehydrated
Media Type D/E Neutralizing Broth
Product Type Microbiology Media
Formulation Pancreatic digest of Casein 5g/L, Yeast extract 2.5g/L, Dextrose 10g/L, Sodium Thioglycollate 1g/L, Sodium Thiosulfate 6g/L, Sodium Bisulfite 2.5g/L, Polysorbate 80 5g/L, Lecithin 7g/L, Bromcresol Purple 0.02g/L
For Use With (Application) Testing of disinfectants and sanitation efficiency
Packaging Type Bottle
Quantity 500 g