missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Cetrimide Agar Base
Specifications
Specifications
| Certifications/Compliance | AOAC, BAM, BS12, CCAM, EP, JP, MCM9, USP |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Cetrimide Agar Base |
| Product Type | Agar |
| Formulation | Pancreatic digest of Gelatin 20g/L, Magnesium Chloride 1.4g/L, Potassium Sulfate 10g/L, Cetrimide (Tetradecyltrimethylammonium Bromide) 0.3g/L, Agar 13.6g/L |
| For Use With (Application) | Selective isolation and identification of P. aeruginosa |
| Packaging Type | Bottle |
| Quantity | 500 g |