missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Buffered Peptone Water Preenrichment

For injured Salmonella in food

$207.26 - $3744.39

Specifications

Certifications/Compliance ISO/DIS, 11014, BAM, CCAM, ISO, USDA
Content And Storage <30°C
Form Dehydrated
Media Type Peptone Water
Product Type Peptone Water
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Type Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Type Price Quantity  
DF1810-08-0
View Documents
BD
218104
10 kg Bottle
Each for $3,744.39
Only null left
 
DF1810-17-9
View Documents
BD
218105
500 g Bottle
Each for $207.26
Only null left
 
Description

Description

Certifications

ISO/DIS, 11014, BAM, CCAM, ISO, USDA

Specifications

Specifications

ISO/DIS, 11014, BAM, CCAM, ISO, USDA
Dehydrated
Peptone Water
Isolation of Salmonella species
<30°C
Peptone Water
Peptone 10g/L, Sodium Chloride 5g/L, Disodium Phosphate 3.5g/L, Monopotassium Phosphate 1.5g/L
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'