missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: B12 Assay Medium

Catalog No. df0457151
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0457151 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0457151 Supplier BD Supplier No. 245710

For use in USP and AOAC methods for microbiological assay of Vitamin B12

Specifications

Certifications/Compliance ISO, 9000
Form Dehydrated
Media Type B12 Assay Medium
Product Type Microbiology Media
Formulation Vitamin assay Casamino Acids 15g/L, Dextrose 40g/L, Asparagine 0.2g/L, Sodium Acetate 20g/L, Ascorbic Acid 4g/L, L-Cystine 0.4g/L, DL-Tryptophan 0.4g/L, Adenine Sulfate 20mg/L, Guanine Hydrochloride 20mg/L, Uracil 20mg/L, Xanthine 20mg/L, Riboflavin 1mg/L, Thiamine Hydrochloride 1mg/L, Biotin 10μg/L, Niacin 2mg/L, p-Aminobenzoic Acid 2mg/L, Calcium Pantothenate 1mg/L, Pyridoxine Hydrochloride 4mg/L, Pyridoxal Hydrochloride 4mg/L, Pyridoxamine Hydrochloride 800μg/L, Folic Acid 200μg/L, Monopotassium Phosphate 1g/L, Dipotassium Phosphate 1g/L, Magnesium Sulfate 0.4g/L, Sodium Chloride 20mg/L, Ferrous Sulfate 20mg/L, Manganese Sulfate 20mg/L, Polysorbate 80 2g/L
For Use With (Application) Isolation of Lactobacillus delbrueckii subspecies Lactis 7830
Packaging Type Bottle
Quantity 100 g