missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Antibiotic Medium 2

Catalog No. DF0270174
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0270174 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0270174 Supplier BD Supplier No. 227020
Only null left

For determining antibiotic potency by the microbiological assay technique

  • Penassay Seed Agar Microbiological assay of antibiotics

Specifications

Certifications/Compliance AOAC, USP
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Antibiotic Medium 2
Product Type Antibiotic Medium
Formulation Beef extract 1.5g/L, Yeast extract 3g/L, Peptone 6g/L, Agar 15g/L
For Use With (Application) Penassay base Agar Microbiological assay of Antibiotics
Packaging Type Bottle
Quantity 500 g