missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Antibiotic Medium 10

Catalog No. DF0463171
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0463171 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0463171 Supplier BD Supplier No. 246310
Edge

For determining antibiotic potency by the microbiological assay technique

  • Antibiotic Medium 10 Polymyxin Base Agar Microbiological assay of antibiotics

Specifications

Certifications/Compliance EP, USP
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Antibiotic Medium 10
Product Type Antibiotic Medium
Formulation Pancreatic digest of Casein 17g/L, Soybean Peptone 3g/L, Dextrose 2.5g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Agar 12g/L, Polysorbate 80 10g/L
For Use With (Application) Antibiotic medium 10 Polymyxin base Agar Microbiological assay of Antibiotics
Packaging Type Bottle
Quantity 500 g