missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: 2x Yeast Extract Tryptone (2xYT) Medium

Catalog No. DF0440170
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0440170 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0440170 Supplier BD Supplier No. B244020
Only null left

For cultivating recombinant strains of Escherichia coli. BD Difco™ Dehydrated Culture Media: 2x Yeast Extract Tryptone (2xYT) Medium is part of a wide selection of dehydrated culture media.

  • Formulation: Pancreatic digest of Casein 16g/L, Yeast extract 10g/L, Yeast extract 5g/L

Order Info

Prices are for units of one each unless otherwise specified.

Specifications

Content And Storage <30°C
Form Dehydrated
Media Type 2XYT
Product Type Microbiology Media
Formulation Pancreatic digest of Casein (Tryptone) 16g/L, Yeast extract 10g/L, NaCl 5g/L
For Use With (Application) Cultivation of recombinant strains of Escherichia coli
Packaging Type Bottle
Quantity 500 g