missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Chromogenic Dehydrated Culture Media: Modified mTEC Agar
Description
Detects degree of water quality deterioration in drinking distribution system enumerates, and monitors the progress in remediation. Provides precise and quantifiable results; also less time consuming and costly than other techniques.
- Any colonies present will exhibit red or magenta color within 24 hours of the test
- Conforms to EPA Method 1603
Order Info
Available in prepared 15 x 60mm plates.
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Light Beige |
| Content And Storage | 2 to 25°C |
| Form | Solid |
| Manufacturing Origin | United States |
| Media Type | Modified m TEC Agar |
| Formulation | No. 3 Proteose Peptone 5g/L, Yeast extract 3g/L, Lactose 10g/L, Sodium Chloride 7.5g/L, Dipotassium Phosphate 3.3g/L, Monopotassium Phosphate 1g/L, Sodium Lauryl Sulfate 0.2g/L, Sodium Desoxycholate 0.1g/L, 5-Bromo-6-chloro-3-indolyl-beta-D-glucuronide 0.5g/L, Agar 15g/L |
| For Use With (Application) | The chromogenic detection and enumeration of thermotolerant Escherichia coli in Water by the membrane filtration technique |
| Packaging Type | Bottle |
| Quantity | 500 g |