Learn More
BD CO2 Gas Generators
For use with Bio-Bag™ Type C Environmental Chambers
Supplier:  BD 261205
Catalog No. 1491033
Only null left
Description
Description
  • Replacement for generator supplied with 14-910-54
  • Can be used with your own enclosure to create a CO2-enriched aerobic environment
  • Each generator contains a crushable ampule that rapidly produces consistent amounts of CO2
  • Use in enclosure (not included) with a volume of at least 340cc
Specifications
Specifications
CO2 Generator
Bio-Bag™
Bulk
Product Suggestions
Product Suggestions